Order Entry
Finland
ContactUsLinkComponent
Eriochrome Black T 1% in sodium chloride Reag. Ph. Eur. 1056801
Eriochrome Black T 1% in sodium chloride Reag. Ph. Eur. 1056801
Catalog # 87873.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Eriochrome Black T 1% in sodium chloride Reag. Ph. Eur. 1056801
Catalog # 87873.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
ADR:9,III
 :461.39 g/mol
EINECS:217-250-3
UN:3077
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:1787-61-7
Formula:C₂₀H₁₂N₃NaO₇S
MDL Number:MFCD00003935