Eriochrome Black T, pure indicator grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1787-61-7
:
Formula:C₂₀H₁₂N₃NaO₇S
MDL Number:MFCD00003935
Merck Index:13,03702
ADR:9,III
:461.39 g/mol
EINECS:217-250-3
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Brown to black Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Suitability:(for compl. det. of metals) Passes test