Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bromocresol green
Bromocresol green 0.1% in ethanol
Bromocresol green 0.1% in ethanol
FFCHFF20710100
:
FF CHEMICALS
:
Bromocresol green 0.1% in ethanol
FFCHFF20710100
:
FF CHEMICALS
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
698.02 g/mol
:
Ambient
:
200-972-8
:
1170
:
3,II
:
GHS02
:
76-60-8
:
C₂₁H₁₄Br₄O₅S
:
MFCD00005874