Bromocresol green sodium salt ACS
:67763-24-0
:62625-32-5
:C₂₁H₁₃Br₄NaO₅S
:MFCD00148898
:230 °C (dec.)
:720 g/mol
:263-657-4
:GHS07
:Ambient
- Appearance:Dark brown to green-black Powder
- Infrared spectrum:Conforms
- Visual transition interval:from pH 3.8 (yellow) to pH 5.4 (blue)
- Clarity of solution:Passes test