Methyl red
Glass bottle, 1 L
FFCHFF0941000
: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- :493-52-7
- :C₁₅H₁₅N₃O₂
- :MFCD00002425
- :269.3 g/mol
- :12,06199
- :178…182 °C
- :1.31 g/cm³ (20 °C)
- :Ambient
- :207-776-1
- Pack type:Glass bottle
- Pk:1 L