Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Congo Red
Congo red
Congo red
CHRO1B-227
:
CHROMA
:
Congo red
CHRO1B-227
:
CHROMA
:
Pk:
25 g
Seq:
3308
:
:
696.67 g/mol
:
12,02562
:
> 360 °C
:
209-358-4
:
GHS08
:
Ambient
:
573-58-0
:
C₃₂H₂₂N₆Na₂O₆S₂
:
MFCD00004028