Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulindac Sulphide
Sulindac Sulphide ≥99% (by TLC)
Sulindac Sulphide ≥99% (by TLC)
Sulindac Sulphide ≥99% (by TLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Sulindac Sulphide ≥99% (by TLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
32004-67-4
:
Formula:
C₂₀H₁₇FO₂S
MDL Number:
MFCD00869764
:
340.4 g/mol
Storage Temperature:
Ambient