Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Novobiocin sodium salt
Novobiocin sodium salt ≥90%
Novobiocin sodium salt ≥90%
:
ENZO LIFE SCIENCES
Novobiocin sodium salt ≥90%
:
ENZO LIFE SCIENCES
:
1476-53-5
:
:
C₃₁H₃₅N₂NaO₁₁
:
MFCD00066541
:
634.614 g/mol