Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zearalenone
Zearalenone ≥99% (by HPLC, TLC)
Zearalenone ≥99% (by HPLC, TLC)
ENZOALX630105M010
:
ENZO LIFE SCIENCES
:
Zearalenone ≥99% (by HPLC, TLC)
ENZOALX630105M010
:
ENZO LIFE SCIENCES
:
Pk:
10 mg
Seq:
2275
Source:
Isolated from Giberella zeae
Purity:
≥99% (HPLC, TLC)
:
:
318.4 g/mol
:
14,10116
:
Freezer
:
241-864-0
:
2811
:
6.1,II
:
GHS06
:
GHS05
:
17924-92-4
:
C₁₈H₂₂O₅