Order Entry
Finland
ContactUsLinkComponent
Diphenylboric acid-2-aminoethyl ester (2-APB) ≥98% (by HPLC)
Diphenylboric acid-2-aminoethyl ester (2-APB) ≥98% (by HPLC)
Catalog # ENZOALX400045M100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Diphenylboric acid-2-aminoethyl ester (2-APB) ≥98% (by HPLC)
Catalog # ENZOALX400045M100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :225.1 g/mol
Storage Temperature:Freezer
EINECS:208-366-5
CAS Number:524-95-8
Formula:C₁₄H₁₆BNO
MDL Number:MFCD00014823