Order Entry
Finland
ContactUsLinkComponent
 
Neomycin sulphate
Catalog # ENZOALX380035G025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Neomycin sulphate
Catalog # ENZOALX380035G025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Merck Index:12,06542
 :908.87 g/mol
Storage Temperature:Ambient
EINECS:215-773-1
CAS Number:1405-10-3
 :MFCD00077833
Formula:C₂₃H₄₆N₆O₁₃·3H₂SO₄
MDL Number:MFCD00150827