Order Entry
Finland
ContactUsLinkComponent
 
Honokiol ≥98% (by HPLC)
  ENZOALX350350M025
 :  
Honokiol ≥98% (by HPLC)
  ENZOALX350350M025
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :266.34 g/mol
 :35354-74-6
 :C₁₈H₁₈O₂
 :MFCD00016674