Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AS601245
AS601245 sum of isomers ≥95% (by HPLC)
AS601245 sum of isomers ≥95% (by HPLC)
AS601245 sum of isomers ≥95% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
AS601245 sum of isomers ≥95% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
345987-15-7
:
Formula:
C₂₀H₁₆N₆S
MDL Number:
MFCD07772196
:
372.45 g/mol