Order Entry
Finland
ContactUsLinkComponent
Fluo-3, pentaammonium salt ≥95% (by HPLC), Ultrapure
Fluo-3, pentaammonium salt ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Fluo-3, pentaammonium salt ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :854.69 g/mol
Storage Temperature:Freezer
CAS Number:339221-91-9