Order Entry
Finland
ContactUsLinkComponent
Acridine Orange 10-nonyl bromide (NAO) ≥95% (by HPLC), Ultrapure
Acridine Orange 10-nonyl bromide (NAO) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52306
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Acridine Orange 10-nonyl bromide (NAO) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52306
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :472.50 g/mol
Storage Temperature:Freezer
CAS Number:75168-11-5
Formula:C₂₆H₃₈BrN₃
MDL Number:MFCD00467605