Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
27-Hydroxycholesterol
27-Hydroxycholesterol ≥98% (by HPLC)
27-Hydroxycholesterol ≥98% (by HPLC)
27-Hydroxycholesterol ≥98% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
27-Hydroxycholesterol ≥98% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
20380-11-4
:
Formula:
C₂₇H₄₆O₂
:
402.65 g/mol
Storage Temperature:
Freezer
Specification Test Results
Purity:
≥98% (HPLC)
Specification Test Results