0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Apicidin
Apicidin ≥99% (by HPLC, TLC)
Apicidin ≥99% (by HPLC, TLC)
ENZOBMLGR3400005
:
ENZO LIFE SCIENCES
:
Apicidin ≥99% (by HPLC, TLC)
ENZOBMLGR3400005
:
ENZO LIFE SCIENCES
:
Pk:
5 mg
Seq:
2210
:
:
623.8 g/mol
:
183506-66-3
:
C₃₄H₄₉N₅O₆