Order Entry
Finland
ContactUsLinkComponent
Despropionylfentanyl ≥99% (by HPLC)
Irritant
Despropionylfentanyl ≥99% (by HPLC)
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Irritant
Despropionylfentanyl ≥99% (by HPLC)
Supplier:  ENZO LIFE SCIENCES

Specification Test Results

  • Source:
    Synthetic
  • Purity:
    ≥99% (HPLC)

Specification Test Results