Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rhodamine 123
Rhodamine 123 ≥95% (by HPLC), Ultrapure
Rhodamine 123 ≥95% (by HPLC), Ultrapure
Rhodamine 123 ≥95% (by HPLC), Ultrapure
ENZOENZ52307
:
ENZO LIFE SCIENCES
:
Rhodamine 123 ≥95% (by HPLC), Ultrapure
ENZOENZ52307
:
ENZO LIFE SCIENCES
:
Pk:
25 mg
Seq:
2315
:
:
380.82 g/mol
:
Freezer
:
62669-70-9
:
C₂₁H₁₇ClN₂O₃
:
MFCD00012664