Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Icilin
Icilin ≥97% (by TLC)
Icilin ≥97% (by TLC)
Irritant
Icilin ≥97% (by TLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Irritant
Icilin ≥97% (by TLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
36945-98-9
:
Formula:
C₁₆H₁₃N₃O₄
MDL Number:
MFCD01689072
:
311.297 g/mol
:
GHS07
Specification Test Results
Purity:
≥97% (TLC)
Specification Test Results