Order Entry
Finland
Orders LinkContactUsLinkComponent
Ciglitazone ≥98%

Ciglitazone ≥98%

Catalog #
Ciglitazone ≥98%
Catalog #
Supplier:  
CAS Number:  74772-77-3
Ciglitazone ≥98%
Ciglitazone ≥98%
CAS Number:  74772-77-3
Product Details & Documents

PPAR ligand

Ciglitazone is a thiazolidinedione type of antihyperglycemic agent. It displays antihyperglycemic activity in genetically obese C57 Bl/6 ob/ob mice and is a selective PPARgamma agonist (EC50=3 µM). Inhibits HUVEC differentiation and angiogenesis and also stimulates adipogenesis and decreases osteoblastogenesis in human mesenchymal stem cells.

Synthetic.

  • CAS Number:74772-77-3
  • Formula:C₁₈H₂₃NO₃S
  • MDL Number:MFCD00865499
  • Molecular Weight:333.446 g/mol