Order Entry
Finland
ContactUsLinkComponent
Luminol (3-Aminophthalhydrazide) ≥95% (by HPLC), CELLESTIAL®, Ultrapure
Luminol (3-Aminophthalhydrazide) ≥95% (by HPLC), CELLESTIAL®, Ultrapure
Catalog # ENZOENZ52354
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Luminol (3-Aminophthalhydrazide) ≥95% (by HPLC), CELLESTIAL®, Ultrapure
Catalog # ENZOENZ52354
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.16 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:521-31-3
Formula:C₈H₇N₃O₂
MDL Number:MFCD00006890