Order Entry
Finland
ContactUsLinkComponent
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98% (by HPLC)
Irritant
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98% (by HPLC)
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Irritant
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98% (by HPLC)
Supplier:  ENZO LIFE SCIENCES

Specification Test Results

  • Purity:
    ≥98% (HPLC)
  • Appearance:
    White to beige to yellow powder

Specification Test Results