Apigenin ≥97% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:520-36-5
:
Formula:C₁₅H₁₀O₅
MDL Number:MFCD00006831
:270.24 g/mol
Frequently Bought Together
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Inoculating loops, Corning® Gosselin™
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Cell culture flasks with standard or filter caps, CELLSTAR®
Supplier: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: Sterilin
Screw caps for micro tubes
Supplier: SARSTEDT
Blotting transfer papers
Supplier: Ahlstrom-Munksjö
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
Cryostorage boxes, KeepIT-100™, WHEATON®
Supplier: DWK Life Sciences
Filter plates, 96-well, for DNA purification, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)