Octaheel Baby Blood Lancets
Supplier: Vitrex Medical A/S
Certificates
About this item
Vitrex Octaheel Baby are sterile cutting safety lancets intended for high volume blood flow with minimum depth of penetration.
- Activation button is secured by safety tab
- Non-detachable safety tab
- Blade is fully shielded before and after use which provides user safety
- Anti-slip texture and one hand operation available for easier handling
- Pre-loaded, button activated design
- Single-use only, sterilised by gamma irradiation
- Ultra sharp blade with high speed cutting securing minimal trauma and fast healing
- Narrowed lancing slot helps for easier positioning
The safety lancets are primarily applicable for newborns and preemies. However, they can also be recommended for other applications, where short penetration depth and/or minimal compression of tissue is important.
Vitrex Octaheel Baby lancet is the latest edition offering improved ergonomy.