Order Entry
Finland
ContactUsLinkComponent
Hydrochloric acid 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504
Hydrochloric acid 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504
Catalog # REGE1043504
Supplier:  REAGECON
CAS Number:  
Hydrochloric acid 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504
Catalog # REGE1043504
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324