Hazardous Goods Bucket
Catalog # RIXIU7S-5101-300-K
Supplier: RIXIUS
Specifications
- Capacity:30 L
- Colour:White
- Description:Hazardous goods bucket, round with lid
- Height:398 mm
- Pk:8
- Ø top/Ø bottom:376/306 mm
Specifications
About this item
UN bucket made of HDPE, white with shell handles, complete with lid type EH300.
- Sturdy plastic bracket
- Ideal bucket for transporting and storing solid hazardous substances
- Airtight and waterproof
- Opaque - protects your contents from light influences
- Odourless and tasteless
- Easy transport even when full
- Weight: 1225 g
Suitable for food. UN - Y approval - suitable for filling goods of packaging group II and III.