Order Entry
Finland
ContactUsLinkComponent
Haematoxylin in aqueous solution according to Delafield for microscopy
Haematoxylin in aqueous solution according to Delafield for microscopy
Catalog # 03971-250ML
CAS Number:  
Haematoxylin in aqueous solution according to Delafield for microscopy
Catalog # 03971-250ML
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :302.28 g/mol
Density:0,950…1,044 g/ml (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:42 °C (closed cup)
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Room Temperature
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111