Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Histopaque®
Histopaque®, sterile filtered, Sigma-Aldrich®
Histopaque®, sterile filtered, Sigma-Aldrich®
Histopaque®, sterile filtered, Sigma-Aldrich®
Supplier:
SIGMA ALDRICH MICROSCOPY
Certificates
Expand image
Histopaque®, sterile filtered, Sigma-Aldrich®
Supplier:
SIGMA ALDRICH MICROSCOPY
Certificates
About this item
:
Density:
1,077 g/ml
:
GHS08
Related Information
Documentation
Certificates
Search by Lot Number
Documentation