Order Entry
Finland
ContactUsLinkComponent
Calibration Cuvettes
  REIC13103410
 :  REICHERT
Calibration Cuvettes
  REIC13103410
 :  REICHERT
  • Description:
    Calibration cuvettes for UNISTAT® bilirubinometer
  • Seq:
    02
  • Pk:
    1
Re-usable glass calibration cuvette with special option coatings assayed at a mid-range Total Serum Bilirubin value. Used to perform an automatic calibration on the Reichert UNISTAT® Bilirubinometer.
Approximate Assay Value 20 mg/dL, 342 µmol/L