Order Entry
Finland
ContactUsLinkComponent
Cerex® RCH OFQC SPE Plate, 96-well
Cerex® RCH OFQC SPE Plate, 96-well
Catalog # TCAT637-0101RCH
Supplier:  Tecan
Cerex® RCH OFQC SPE Plate, 96-well
Catalog # TCAT637-0101RCH
Supplier:  Tecan
Specifications
  • No. of wells:
    96
  • Description:
    Plate RCH Cerex® OFQC 1 ml
  • Phase:
    OFQC
  • Filling:
    10 mg
  • Pk:
    1
Specifications
About this item
Proprieatary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
  • Ion-exchange
  • Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.