Order Entry
Finland
ContactUsLinkComponent
 
Sodium hypochlorite (3.5% Cl₂) in aqueous solution
  FFCHFF4050500
 :  
Sodium hypochlorite (3.5% Cl₂) in aqueous solution
  FFCHFF4050500
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

 :
 :74.44 g/mol
 :Refrigerator
 :231-668-3
 :7681-52-9
 :ClNaO
 :MFCD00011120