(±)-β-Citronellol
Glass bottle, 100 mL
Catalog # ROTH6220.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
- CAS Number:106-22-9
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00002935
- Molecular Weight:156.27 g/mol
- Merck Index:13,02354
- Boiling Pt:222 °C (1013 hPa)
- Density:0.86 g/cm³ (20 °C)
- Flash Pt:100 °C
- EINECS:203-375-0
- UN:3082
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



