ESD Safety Shoes, Low, LUAN, S1PS
Supplier: ELTEN
Certificates
About this item
Mesh material with recycled material content/Microfibre with recycled material content.
- Breathable lining with recycled material content
- Padded tongue
- Full-length inlay sole SPORTIVE ESD grey with recycled material content
- Non-metallic, penetration-resistant midsolewith recycled material content
- TPU/PU sole MAXXIMO
- Steel toe cap
Safety shoes are designed for use in areas where there is a risk of electrostatic discharge (ESDS/ESD) and workplaces on hard undergrounds. The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides, creating more energy in every step.
Certifications: EN ISO 20345 S1PS FO/SR, form A.
Specifications
- Catalog No:
- ELTN729121/47
- ELTN729121/46
- ELTN729121/45
- ELTN729121/39
- ELTN729121/44
- ELTN729121/40
- ELTN729121/36
- ELTN729121/37
- ELTN729121/43
- ELTN729121/35
- ELTN729121/38
- ELTN729121/42
- ELTN729121/41
- ELTN729121/48
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001