Heat Transfern Liquid, Ultra 301
Catalog # LAUDLZB353
Supplier: LAUDA
CAS Number:
Specifications
- Pk:20 L
- Description:Heat transfern liquid, ultra 301
- Density:858 kg/m³ at 20 °C
- Flash point:245 °C
- Viscosity:76.5 mm²/s at 20 °C
Specifications
About this item
Heat transfer liquid for temperatures up to over 300 °C.
- Silicone-free liquid, ideal for surface-sensitive applications
- The safety data sheet must be observed for uses in bath thermostats