Order Entry
Finland
ContactUsLinkComponent
Flame-Resistant Anti-Static Balaclava
Flame-Resistant Anti-Static Balaclava
Catalog # PRTWFR18BKR
Supplier:  Portwest
Flame-Resistant Anti-Static Balaclava
Catalog # PRTWFR18BKR
Supplier:  Portwest
Specifications
  • Colour:
    Black
  • Pk:
    1
Specifications
About this item
Ideal for cooler conditions to keep the head and neck warm.
  • Inherent flame-resistant qualities will not diminish with washing
  • Protection against radiant, convective, and contact heat
  • Anti-static
  • High cotton content for superior comfort
The 2-layer ribbed fabric allows for a snug and comfortable fit.
Certifications: EN 1149 -5. CE-CAT III.