Tripod rods, square, PASS
Supplier: PHYWE
Certificates
About this item
Tripod rods with square cross section 12×12 mm. Made of galvanized galvanized steel pipe. One end of the rod is provided with a cylindrical rod (50×10 mm). The other end closed. Square rods provide unmatched torsional rigidity compared to cylindrical frame rods.