Hole siren
Catalog # 765-0126
Supplier: CORNELSEN EXPERIMENTA
Specifications
- Description:Hole siren
- Pk:1
Specifications
About this item
Metal plate with four rows of holes at different distances. By simultaneously setting the disc in rotation and blowing with a blowpipe, a major chord is obtained. The disc is used together with a hand-driven rotary apparatus or an electric motor. Disc Ø: 200 mm.