Tambourine
765-0157
- Description:Tambourine
- Pk:1
Metal frame with tension spring and membrane of Papyrolin. With the help of two tambourines and a pendulum ball, experiments with pressure waves can be performed.
- Diameter: 190 mm
Metal frame with tension spring and membrane of Papyrolin. With the help of two tambourines and a pendulum ball, experiments with pressure waves can be performed.