Order Entry
Finland
ContactUsLinkComponent
Trolley to inclined plane
Trolley to inclined plane
Catalog # 765-0925
undefined
Trolley to inclined plane
Catalog # 765-0925
Specifications
  • Description:
    Spare wheels
  • Pk:
    1
Specifications
About this item
Trolley, 100 g, of lacquered sheet metal with nylon wheels. Equipped with hook and hole for mounting weights.
  • Dimensions: 110 × 50 × 45 mm