Order Entry
Finland
ContactUsLinkComponent
Inclined plane with roller
Inclined plane with roller
Catalog # 765-1379
Supplier:  PHYWE
undefined
Inclined plane with roller
Catalog # 765-1379
Supplier:  PHYWE

Specifications

  • Description:
    Inclined plane with roller
  • Pk:
    1

Specifications

About this item

Practical device for demonstration and determination of the force that keeps a body in balance on an inclined plane.

  • Quick installation without special settings
  • The angle of the web is freely adjustable between 15 and 45 °
  • Roll as a force object, forces that affect meet in the center of gravity, no moments
  • Two demonstration rulers and an angular scale for geometric determinations
  • Three holders for dynamometers
  • The dynamometers can be attached parallel, perpendicularly or horizontally to the track
  • Clear demonstration of force analysis with force parallelogram
  • Power components perpendicular and within a measuring range between 105 ° and 135 °

Necessary accessories: 2 Dynamometers (765-1168).