Order Entry
Finland
Orders LinkContactUsLinkComponent
Material samples
Material samples
  765-0115
Marketsource
Material samples
  765-0115
Marketsource
Material samples
Material samples
Material samples
1 * 15 Items
Set of 15 samples for examination of mechanical, calorimetric, magnetic and electrical properties. Materials: Aluminum, Iron, Glass, Rubber, Wood, Porcelain, Copper, Nickel, Cardboard, Woolen fabric, Coal, Foam, Zinc, PVC, AlNiCo.
  • Delivered in a plastic box
  • Description:
    Material samples