Order Entry
Finland
ContactUsLinkComponent
Hydrostatic paradox
Hydrostatic paradox
Catalog # 765-0138
undefined
Hydrostatic paradox
Catalog # 765-0138
Specifications
  • Description:
    Hydrostatic paradox
  • Pk:
    1
Specifications
About this item
Shows that the bottom pressure in a liquid depends only on the height and not the shape of the container. The fluid pressure is transferred via a rubber diaphragm to an adjustable hand. Comes with four containers of different shapes. Dimensions.
  • Height: 290 mm
  • Bottom plate: 235 × 130 mm