Diver, Cartesian
Cartesian diver
Catalog # 765-1178
Marketsource
Supplier: PHYWE
Jump to:
Product Details & Documents
Divers of glass, placed in a bottle filled with water. If the liquid pressure increases, water flows into the diver, whose average density increases. This results in the diver sinking to rise again towards the surface when the pressure is reduced.
Specifications
- Description:Cartesian diver