Magdeburger hemisphere
Catalog # 765-0147
Supplier: CORNELSEN EXPERIMENTA
Specifications
- Description:Magdeburger hemisphere
- Pk:2
Specifications
About this item
Simpler model of Magdeburger hemisphere. Equipped with folding hooks with which the air is evacuated.
- Dimensions: 40 mm