Order Entry
Finland
ContactUsLinkComponent
Metal cylinders, same mass
Metal cylinders, same mass
Catalog # 765-0114
undefined
Metal cylinders, same mass
Catalog # 765-0114
Specifications
  • Description:
    Metal cylinders, same mass
  • Pk:
    1
Specifications
About this item
Rate of 6 pcs. metal cylinders of different materials. Al, Cu, Fe, Ms, Pb, and Sn. The cylinders have the same mass but different volume.