Headband with elderflower balls
Catalog # 765-0923
Supplier: GR NNA PLASTEMBALLAGE AB
Specifications
- Description:Headband with elderflower balls
- Pk:1
Specifications
About this item
Ribbons with elderflower balls suspended in thin threads. Placed around the head of a person who stands on an insulating surface and can be charged with the help of a belt generator. The balls stand out correctly. The belt can also be placed directly on the belt of the belt generator.