Tissue paper screen
Tissue paper screen
Catalog # 765-0190
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
When charging, the screen unfolds. Equipped with 4 mm rod suitable for influence machine, belt generator and Faraday's cage.
- Length: 120 mm
Specifications
- Description:Tissue paper screen