Order Entry
Finland
ContactUsLinkComponent
Plexiglas tube with ball
Plexiglas tube with ball
Catalog # 765-0188
undefined
Plexiglas tube with ball
Catalog # 765-0188
Specifications
  • Description:
    Plexiglas tube with ball
  • Pk:
    1
Specifications
About this item
Apparatus for demonstrating the movement of a charged body. Consists of plexiglass tubes with two metal electrodes, ball with metallic surface and 4 mm connection sleeves. When connected to a high voltage source, the ball moves back and forth between the electrodes.
  • Length: 150 mm
  • Diam, ball: 43 mm