Order Entry
Finland
Orders LinkContactUsLinkComponent
Conductor balls
Conductor balls
Marketsource
Supplier:   PHYWE
Conductor balls
Supplier:  PHYWE
Marketsource
Certificates
Conductor balls
3 Options Available Below
Product Details & Documents
For electrostatic experiments, for example, determination of the charge of a sphere and induction experiments. Hollow surface-treated brass sphere equipped with 4 mm holes for connecting banana connectors.
Product Family Options
Product Information
  • Availability
    Price
  • 765-1190
    Conductor ball, diameter 20 mm
    Conductor ball, diameter 20 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 97,50 €
    97,50 €
    1 * 1 Each
       97,50 €
    Specifications:
    Marketsource
    Marketsource
  • 765-1191
    Conductor ball, diameter 40 mm
    Conductor ball, diameter 40 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 160,00 €
    160,00 €
    1 * 1 Each
       160,00 €
    Specifications:
    Marketsource
    Marketsource
  • 765-1192
    Conductor ball, diameter 120 mm
    Conductor ball, diameter 120 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 484,00 €
    484,00 €
    1 * 1 Each
       484,00 €
    Specifications:
    Marketsource
    Marketsource